Assay Details
| Gene Symbol: LOC100072199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005602032.3 | XP_005602089.2 | 251 | 155 | ||
| XM_023623856.1 | XP_023479624.1 | 251 | 155 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005602032.3, XM_023623856.1 transcripts.
Set Membership: |
|

LOC100072199
Genomic Map