Assay Details
| Gene Symbol: FKBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603761.3 | XP_005603818.1 | 113 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005603761.3 transcripts.
Set Membership: |
|

FKBPL
Genomic Map