Assay Details
| Gene Symbol: LOC100050760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001491140.6 | XP_001491190.3 | 85 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001491140.6 transcripts.
Set Membership: |
|

LOC100050760
Genomic Map