Assay Details
| Gene Symbol: ANKRD61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362687.3 | XP_003362735.1 | 1222 | 72 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD61| Gene Symbol: ANKRD61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362687.3 | XP_003362735.1 | 1222 | 72 | ||