Assay Details
| Gene Symbol: ANKRD61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362687.3 | XP_003362735.1 | 1315 | 85 | ||
| XM_023655257.1 | XP_023511025.1 | 1312 | 85 | |||
More Information
Set Membership: |
|

ANKRD61
Genomic Map