Assay Details
| Gene Symbol: TEDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599055.3 | XP_005599112.1 | 175 | 112 | ||
| XM_005599056.3 | XP_005599113.1 | 175 | 112 | |||
| XM_005599057.3 | XP_005599114.1 | 175 | 112 | |||
| XM_014730233.2 | XP_014585719.1 | 175 | 112 | |||
More Information
Set Membership: |
|

TEDC2
Genomic Map