Assay Details
| Gene Symbol: C21H19orf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604103.3 | XP_005604160.1 | 203 | 83 | ||
| XM_014734757.2 | XP_014590243.1 | 202 | 83 | |||
| XM_014734758.2 | XP_014590244.1 | 200 | 83 | |||
| XM_014734759.2 | XP_014590245.1 | 201 | 83 | |||
| XM_023625372.1 | XP_023481140.1 | 200 | 83 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005604103.3, XM_014734757.2, XM_014734758.2, XM_014734759.2, XM_023625372.1 transcripts.
Set Membership: |
|

C21H19orf44
Genomic Map