Assay Details
| Gene Symbol: C21H19orf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604103.3 | XP_005604160.1 | 1280 | 70 | ||
| XM_014734757.2 | XP_014590243.1 | 1279 | 70 | |||
| XM_014734758.2 | XP_014590244.1 | 1277 | 70 | |||
| XM_014734759.2 | XP_014590245.1 | 1278 | 70 | |||
| XM_023625371.1 | XP_023481139.1 | 1229 | 70 | |||
| XM_023625372.1 | XP_023481140.1 | 1277 | 70 | |||
More Information
Set Membership: |
|

C21H19orf44
Genomic Map