Assay Details
| Gene Symbol: C21H19orf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604103.3 | XP_005604160.1 | 1775 | 90 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C21H19orf44| Gene Symbol: C21H19orf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604103.3 | XP_005604160.1 | 1775 | 90 | ||