Assay Details
| Gene Symbol: BORCS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604142.3 | XP_005604199.1 | 406 | 89 | ||
| XM_005604143.3 | XP_005604200.1 | 406 | 89 | |||
| XM_014734798.2 | XP_014590284.1 | 406 | 89 | |||
More Information
Set Membership: |
|

BORCS8
Genomic Map