Assay Details
| Gene Symbol: LOC100055638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014727809.2 | XP_014583295.2 | 1151 | 157 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100055638| Gene Symbol: LOC100055638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014727809.2 | XP_014583295.2 | 1151 | 157 | ||
Set Membership: |
|