Assay Details
| Gene Symbol: GNG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729628.2 | XP_014585114.1 | 528 | 120 | ||
| XM_014729629.2 | XP_014585115.1 | 439 | 120 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GNG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729628.2 | XP_014585114.1 | 528 | 120 | ||
| XM_014729629.2 | XP_014585115.1 | 439 | 120 | |||
Set Membership: |
|