Assay Details
| Gene Symbol: LOC100054420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014732031.1 | XP_014587517.1 | 502 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100054420| Gene Symbol: LOC100054420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014732031.1 | XP_014587517.1 | 502 | 97 | ||
Set Membership: |
|