Assay Details
| Gene Symbol: B3GALT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014734347.2 | XP_014589833.1 | 675 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014734347.2 transcripts.
Set Membership: |
|

B3GALT4
Genomic Map