Assay Details
| Gene Symbol: LOC100071129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014734833.2 | XP_014590319.2 | 189 | 68 | ||
| XM_023625537.1 | XP_023481305.1 | 921 | 68 | |||
More Information
Set Membership: |
|

LOC100071129
Genomic Map