Assay Details
| Gene Symbol: CD300E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738912.2 | XP_014594398.1 | 1179 | 60 | ||
| XM_023652086.1 | XP_023507854.1 | 1538 | 60 | |||
| XM_023652087.1 | XP_023507855.1 | 1538 | 60 | |||
More Information
Set Membership: |
|

CD300E
Genomic Map