Assay Details
| Gene Symbol: EFCAB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738947.2 | XP_014594433.2 | 156 | 94 | ||
| XM_023638857.1 | XP_023494625.1 | 156 | 94 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EFCAB10| Gene Symbol: EFCAB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738947.2 | XP_014594433.2 | 156 | 94 | ||
| XM_023638857.1 | XP_023494625.1 | 156 | 94 | |||
Set Membership: |
|