Assay Details
Gene Symbol: TAS2R7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014740555.1 | XP_014596041.1 | 621 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
