Assay Details
| Gene Symbol: LOC100630860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014741556.2 | XP_014597042.1 | 65 | 117 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100630860| Gene Symbol: LOC100630860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014741556.2 | XP_014597042.1 | 65 | 117 | ||
Set Membership: |
|