Assay Details
| Gene Symbol: SDR9C7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488339.5 | XP_001488389.2 | 5393 | 68 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SDR9C7| Gene Symbol: SDR9C7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488339.5 | XP_001488389.2 | 5393 | 68 | ||
Set Membership: |
|