Assay Details
| Gene Symbol: INSYN2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488676.4 | XP_001488726.2 | 1858 | 118 | ||
| XM_023637117.1 | XP_023492885.1 | 1502 | 118 | |||
More Information
Set Membership: |
|

INSYN2A
Genomic Map