Assay Details
| Gene Symbol: LOC100055175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489482.2 | XP_001489532.2 | 907 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100055175| Gene Symbol: LOC100055175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489482.2 | XP_001489532.2 | 907 | 81 | ||
Set Membership: |
|