Assay Details
| Gene Symbol: LOC111767619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023616853.1 | XP_023472621.1 | 270 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023616853.1 transcripts.
Set Membership: |
|

LOC111767619
Genomic Map