Assay Details
| Gene Symbol: MRPL57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023621372.1 | XP_023477140.1 | 155 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023621372.1 transcripts.
Set Membership: |
|

MRPL57
Genomic Map