Assay Details
| Gene Symbol: CDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001491903.5 | XP_001491953.2 | 247 | 101 | ||
| XM_005597141.2 | XP_005597198.1 | 807 | 101 | |||
| XM_023651992.1 | XP_023507760.1 | 391 | 101 | |||
More Information
Set Membership: |
|

CDK3
Genomic Map