Assay Details
| Gene Symbol: RBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494823.2 | XP_001494873.1 | 5049 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494823.2 | XP_001494873.1 | 5049 | 77 | ||
Set Membership: |
|