Assay Details
| Gene Symbol: LOC100063838 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494931.5 | XP_001494981.3 | 627 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100063838| Gene Symbol: LOC100063838 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494931.5 | XP_001494981.3 | 627 | 88 | ||