Assay Details
| Gene Symbol: LOC111768656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023621908.1 | XP_023477676.1 | 246 | 75 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC111768656| Gene Symbol: LOC111768656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023621908.1 | XP_023477676.1 | 246 | 75 | ||