Assay Details
| Gene Symbol: LOC100629418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625069.1 | XP_023480837.1 | 309 | 74 | ||
| XM_023625072.1 | XP_023480840.1 | 315 | 74 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023625069.1, XM_023625072.1 transcripts.
Set Membership: |
|

LOC100629418
Genomic Map