Assay Details
| Gene Symbol: LOC100629418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625069.1 | XP_023480837.1 | 916 | 92 | ||
| XM_023625071.1 | XP_023480839.1 | 705 | 92 | |||
More Information
Set Membership: |
|

LOC100629418
Genomic Map