Assay Details
| Gene Symbol: LOC111769620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625322.1 | XP_023481090.1 | 1146 | 72 | ||
| XM_023625323.1 | XP_023481091.1 | 1227 | 72 | |||
| XM_023625326.1 | XP_023481094.1 | 1835 | 72 | |||
More Information
Set Membership: |
|

LOC111769620
Genomic Map