Assay Details
| Gene Symbol: TNFRSF6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023626953.1 | XP_023482721.1 | 791 | 89 | ||
| XM_023626954.1 | XP_023482722.1 | 791 | 89 | |||
More Information
Set Membership: |
|

TNFRSF6B
Genomic Map