Assay Details
| Gene Symbol: LIME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023626955.1 | XP_023482723.1 | 552 | 99 | ||
| XM_023626956.1 | XP_023482724.1 | 670 | 99 | |||
| XM_023626957.1 | XP_023482725.1 | 470 | 99 | |||
More Information
Set Membership: |
|

LIME1
Genomic Map