Assay Details
| Gene Symbol: DYNC2I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023629052.1 | XP_023484820.1 | 447 | 67 | ||
| XM_023629053.1 | XP_023484821.1 | 447 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DYNC2I2| Gene Symbol: DYNC2I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023629052.1 | XP_023484820.1 | 447 | 67 | ||
| XM_023629053.1 | XP_023484821.1 | 447 | 67 | |||
Set Membership: |
|