Assay Details
| Gene Symbol: SPATA2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023637541.1 | XP_023493309.1 | 496 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023637541.1 transcripts.
Set Membership: |
|

SPATA2L
Genomic Map