Assay Details
| Gene Symbol: PRDM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023637592.1 | XP_023493360.1 | 439 | 79 | ||
| XM_023637593.1 | XP_023493361.1 | 439 | 79 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRDM7| Gene Symbol: PRDM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023637592.1 | XP_023493360.1 | 439 | 79 | ||
| XM_023637593.1 | XP_023493361.1 | 439 | 79 | |||
Set Membership: |
|