Assay Details
| Gene Symbol: PRDM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023637592.1 | XP_023493360.1 | 1194 | 107 | ||
| XM_023637593.1 | XP_023493361.1 | 1194 | 107 | |||
More Information
Set Membership: |
|

PRDM7
Genomic Map