Assay Details
| Gene Symbol: C3H4orf48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638509.1 | XP_023494277.1 | 304 | 127 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C3H4orf48| Gene Symbol: C3H4orf48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638509.1 | XP_023494277.1 | 304 | 127 | ||
Set Membership: |
|