Assay Details
| Gene Symbol: LOC100056976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638808.1 | XP_023494576.1 | 1791 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100056976| Gene Symbol: LOC100056976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638808.1 | XP_023494576.1 | 1791 | 80 | ||