Assay Details
| Gene Symbol: LOC100060536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023641520.1 | XP_023497288.1 | 1851 | 74 | ||
| XM_023641521.1 | XP_023497289.1 | 1763 | 74 | |||
More Information
Set Membership: |
|

LOC100060536
Genomic Map