Assay Details
| Gene Symbol: LOC100147270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023642047.1 | XP_023497815.1 | 617 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100147270| Gene Symbol: LOC100147270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023642047.1 | XP_023497815.1 | 617 | 71 | ||