Assay Details
| Gene Symbol: SPSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023643057.1 | XP_023498825.1 | 956 | 67 | ||
| XM_023643058.1 | XP_023498826.1 | 945 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPSB2| Gene Symbol: SPSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023643057.1 | XP_023498825.1 | 956 | 67 | ||
| XM_023643058.1 | XP_023498826.1 | 945 | 67 | |||
Set Membership: |
|