Assay Details
| Gene Symbol: LOC100072895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646232.1 | XP_023502000.1 | 662 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100072895| Gene Symbol: LOC100072895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646232.1 | XP_023502000.1 | 662 | 95 | ||
Set Membership: |
|