Assay Details
| Gene Symbol: CABP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646881.1 | XP_023502649.1 | 405 | 72 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CABP7| Gene Symbol: CABP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646881.1 | XP_023502649.1 | 405 | 72 | ||