Assay Details
Gene Symbol: ZNF516-DT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648278.1 | XP_023504046.1 | 74 | 110 |

More Information
Set Membership: |
![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF516-DT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648278.1 | XP_023504046.1 | 74 | 110 |
Set Membership: |
![]() ![]() |