Assay Details
| Gene Symbol: PXDNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023648330.1 | XP_023504098.1 | 985 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PXDNL| Gene Symbol: PXDNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023648330.1 | XP_023504098.1 | 985 | 91 | ||
Set Membership: |
|