Assay Details
| Gene Symbol: PYCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023649121.1 | XP_023504889.1 | 142 | 79 | ||
| XM_023649122.1 | XP_023504890.1 | 142 | 79 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PYCR3| Gene Symbol: PYCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023649121.1 | XP_023504889.1 | 142 | 79 | ||
| XM_023649122.1 | XP_023504890.1 | 142 | 79 | |||
Set Membership: |
|