Assay Details
| Gene Symbol: CCNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023649746.1 | XP_023505514.1 | 4111 | 123 | ||
| XM_023649748.1 | XP_023505516.1 | 4111 | 123 | |||
More Information
Set Membership: |
|

CCNP
Genomic Map