Assay Details
| Gene Symbol: IL4I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650192.1 | XP_023505960.1 | 480 | 102 | ||
| XM_023650193.1 | XP_023505961.1 | 421 | 102 | |||
| XM_023650196.1 | XP_023505964.1 | 269 | 102 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023650196.1 transcripts.
Set Membership: |
|

IL4I1
Genomic Map