Assay Details
| Gene Symbol: IL4I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650192.1 | XP_023505960.1 | 830 | 99 | ||
| XM_023650193.1 | XP_023505961.1 | 771 | 99 | |||
| XM_023650194.1 | XP_023505962.1 | 736 | 99 | |||
| XM_023650195.1 | XP_023505963.1 | 795 | 99 | |||
| XM_023650196.1 | XP_023505964.1 | 619 | 99 | |||
More Information
Set Membership: |
|

IL4I1
Genomic Map